MQuant™ pH Test Strips and Indicator Papers
MQuant™ pH Test Strips and Indicator Papers
Actual product may vary
MilliporeSigma
MQuant™ pH Test Strips and Indicator Papers
Measure pH onsite without instruments or extensive preparation
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH RangeTypeQuantity
FISM10953500010 to 14Universal pH Indicator Strip100 Strips
FISM10953200017.5 to 14Acilit pH Indicator Strip100 Strips in Plastic Box
Catalog number FISM1095350001
Price (SGD)
78.00
EA
Add to cart
pH Range:
0 to 14
Type:
Universal pH Indicator Strip
Quantity:
100 Strips
Price (SGD)
78.00
EA
Add to cart
Fit into a pocket and provide easy, rapid results—suited to all media in environmental analysis and industrial in-process controls.
  • Differentiated color gradations for a more precise evaluation
  • Traditional indicator paper or MQuant™ nonbleeding universal strips available
Traditional strips
  • High-quality papers impregnated with indicator or mixed-indicator solutions, cut to size and dried
  • Supplied in a roll format that protects strips against moisture, light or ambient gases for longer storage life
Plastic indicator strips provide differentiated color gradations for a more precise evaluation
  • Special indicator dyes covalently bonded to reagent papers
  • Indicator does not bleed, so strips will not contaminate the medium, even if left for long periods of time
Long shelf life
  • Can be used for three to five years when stored at 10 to 25°C in a light-protected area
  • Reseal quickly after each use
Consistently high quality
  • Tests checked and calibrated using certified buffer solutions
  • Buffer solutions are traceable to primary reference materials from NIST™ and PTB
Specifications
pH Range0 to 14
For Use With (Application)Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination
Graduations15
Quantity100 Strips
Shelf Life3 to 5 Years
Storage Requirements10°C to 25°C
TypeUniversal pH Indicator Strip
Unit SizeEA