Check Order Status
TISAB II, For Fluoride, Certified, LabChem™
Element Search
Structure Search
Fisherbrand
TISAB II, For Fluoride, Certified, LabChem™
Have Questions?
Catalog Number
Quantity
FISLC261304
4 L
Catalog number
FISLC261304
Price (SGD)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 64-19-7, 1310-73-2, 125572-95-4
Specifications
Chemical Name or Material
TISAB II
Color
Colorless
Name Note
For Fluoride
Health Hazard 1
Warning
CAS Max %
83.7
View more