Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Potassium Chloride Saturated, Electrode Filling Solution, Certified, ≥ 4.0M, LabChem™
Have Questions?
Catalog NumberQuantity
FISLC188001500 mL
Catalog number FISLC188001
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium Chloride Saturated
ColorColorless
Name NoteElectrode Filling Solution
CAS7447-40-7
CAS Max %73.6
View more