Check Order Status
Buffer Solution pH 13.0 at 25°C, Certified, LabChem™
Element Search
Structure Search
Fisher Catalogue
Buffer Solution pH 13.0 at 25°C, Certified, LabChem™
Have Questions?
Catalog Number
Quantity
Packaging
FISLC125651
500 mL
Poly Bottle
Catalog number
FISLC125651
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 1310-73-2, 89-83-8
Specifications
Chemical Name or Material
Buffer Solution pH 13.0 at 25°C
Health Hazard 1
Danger
CAS Max %
99.24
Color
Brown to Colorless
Health Hazard 2
GHS H Statement