Kimble™ Kontes™ Ultra-Ware™ HPLC Reservoirs
Kimble™ Kontes™ Ultra-Ware™ HPLC Reservoirs
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Ultra-Ware™ HPLC Reservoirs
Designed for the preparation, storage, and delivery of liquid chromatography mobile phases
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Bottom Shape
FISK953901-200320,000 mLConical
FISK953901-100310,000 mLConical
Catalog number FISK953901-2003
Price (SGD)
-
Capacity (Metric):
20,000 mL
Bottom Shape:
Conical
  • Manufactured from Type I, Class A borosilicate glass with external plastic safety coating
  • Safety coating has UV-blocking agent that blocks light transmittance up to 385nm, preventing photodegradation of light-sensitive solvents
  • Screw thread: GL45-4
  • Maximum temperature: 120°C
  • Available in flat or conical bottoms; conical-bottom reservoirs deliver virtually all of the mobile phase without having to tilt reservoir and risk solvent spill
Specifications
Certifications/ComplianceUSP Type I and ASTM E438, Type I, Class A
CoatingPVC
Diameter (English) Outer12.4 in.
Capacity (Metric)20,000 mL
MaterialBorosilicate Glass
TypeHPLC Reservoir
For Use With (Application)Designed for the preparation, storage, and delivery of liquid chromatography mobile phases
Height (English)22.75 in.
Height (Metric)578 mm
IncludesGL45-4 screw thread
Max. Temperature (English)248°F
Max. Temperature (Metric)120°C
Diameter (Metric) Outer315 mm
Bottom ShapeConical
Unit SizeEA