Kimble™ Kontes™ Filtration Assemblies: Flasks With Inner Joint 40/35, for 90mm Glass Assemblies
Kimble™ Kontes™ Filtration Assemblies: Flasks With Inner Joint 40/35, for 90mm Glass Assemblies
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Filtration Assemblies: Flasks With Inner Joint 40/35, for 90mm Glass Assemblies
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Inner Joint 40/35, for 90mm Glass Assemblies, Borosilicate Glass, Description: Funnel for 47mm assemblies, For Use With: Ultra-Ware 90mm all-glass Microfiltration Assembly, Standard Taper Joint: 40/35
Have Questions?
Catalog NumberVolume (Metric)
FISK953829-00004000 mL
Catalog number FISK953829-0000
Price (SGD)
1,206.00
EA
Add to cart
Volume (Metric):
4000 mL
Price (SGD)
1,206.00
EA
Add to cart
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Inner Joint 40/35, for 90mm Glass Assemblies, Borosilicate Glass, Description: Funnel for 47mm assemblies, For Use With: Ultra-Ware 90mm all-glass Microfiltration Assembly, Standard Taper Joint: 40/35
Specifications
For Use With (Equipment)Ultra-Ware 90 mm all-glass Microfiltration Assembly
MaterialBorosilicate Glass
Standard Taper Joint40/35
TypeFlask
Volume (Metric)4000 mL
Unit SizeEA