Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Stainless Steel Support and GL 45 Style Bottle (47mm)
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Stainless Steel Support and GL 45 Style Bottle (47mm)
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Stainless Steel Support and GL 45 Style Bottle (47mm)
Designed to allow connection of a 47 mm or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
Have Questions?
Catalog number FISK953800-5347
Price (SGD)
1,955.00
EA
Add to cart
Price (SGD)
1,955.00
EA
Add to cart
It has a stainless steel support and is recommended for filtering viscous or proteinaceous solutions or to produce ultra-clean filtrate.
  • Supplied with funnel, clamp, 304 stainless steel support screen, PTFE gasket, glass support base, tubing adapter with o-ring, PBT cap, sealing ring and vacuum/pressure bottle
  • Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
  • All components are steam autoclavable
  • 80 x 80 304 stainless steel mesh, 0.0055 in. opening size, 19.4% open area, 0.007 in. wire diameter
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Note:
For replacement Hose Barb Connector with O-Ring, see 7364001413.
Specifications
MaterialStainless Steel (316)
TypeMicrofiltration Assembly
Volume (Metric)300 mL
Width (English)4 in.
Width (Metric)10.2 cm
Height (English)14.5 in.
Height (Metric)36.8 cm
Unit SizeEA