Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Have Questions?
Catalog NumberCapacity (Metric)
FISK953760-10021000 mL
Catalog number FISK953760-1002
Price (SGD)
571.00
EA
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
571.00
EA
Add to cart
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
  • KimCote™ plastic safety coating
  • ≠2 side arm for connection to 0..25 in. (6mm) I.D. flexible tubing
  • 125mL flask: With No. 5 stopper joint
  • 1, 2 and 4L flasks: With No. 8 stopper joint
Specifications
Diameter (Metric) Inner Tubing0.93 cm
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Closure Size8
Unit SizeEA