Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (47mm)
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (47mm)
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (47mm)
Used for general filtration and is designed to handle up to 500mL of sample liquids for the analysis of particulate or microbiological contamination
Have Questions?
Catalog number FISK953755-0000
Price (SGD)
832.00
CS
Add to cart
Price (SGD)
832.00
CS
Add to cart
  • Supplied with a 40-60 micron porosity fritted glass support base, a 300mL graduated funnel, an anodized aluminum clamp and a #8 silicone stopper
  • Funnels are graduated from 100 to 250mL in 25mL increments
  • Prefilter size is 35mm
  • Approximate filter area is 9.6cm2
  • Connection to 1 liter filtration flask (available as an accessory) is made with a #8 silicone stopper
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
MaterialGlass
IncludesFritted glass support base, 300mL graduated funnel, anodized aluminum clamp and a #8 silicone stopper
Volume (Metric)300 mL
Width (English)3 in.
Width (Metric)7.6 cm
Height (English)9 in.
Height (Metric)22.9 cm
Unit SizeCS