Kimble™ Microfiltration Assembly Hardware
Kimble™ Microfiltration Assembly Hardware
Actual product may vary
Kimble™ Microfiltration Assembly Hardware
Kimble™ Microfiltration Assembly Hardware
DWK Life Sciences
Kimble™ Microfiltration Assembly Hardware
Use Kimble™ Microfiltration Assembly Hardware when filtering large volumes of viscous or proteinaceous solutions to give the maximum flow rate.
Have Questions?
Catalog NumberMaterialType
FISK953752-0001Fritted GlassGlass Support
Catalog number FISK953752-0001
Price (SGD)
505.00
EA
Add to cart
Material:
Fritted Glass
Type:
Glass Support
Price (SGD)
505.00
EA
Add to cart
  • Microfiltrational assembly 1000mL
  • Funnel, 4000mL
  • Flask, borosilicate glass, 47mm
  • Glass support and ground joint 90mm
  • Anodized aluminum clamp and filtration flask
Specifications
For Use With (Equipment)Ultra-Ware 47mm Microfiltration Assembly
MaterialFritted Glass
TypeGlass Support
Length (Metric)47 mm
Unit SizeEA