Check Order Status
Kimble™ Kontes™ Parts for Microfiltration Assemblies
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Parts for Microfiltration Assemblies
For use with ULTRA-WARE™ microfiltration assembly
Have Questions?
Catalog Number
Material
Type
Diameter (Metric) Disc
Volume (Metric)
FISK953751-0000
Glass
Funnel
47 mm
300 mL
Catalog number
FISK953751-0000
Price (SGD)
338.00
EA
Add to cart
Material:
Glass
Type:
Funnel
Diameter (Metric) Disc:
47 mm
Volume (Metric):
300 mL
Price (SGD)
338.00
EA
Add to cart
For use with ULTRA-WARE™ microfiltration assembly
Specifications
Material
Glass
For Use With (Application)
ULTRA-WARE microfiltration assembly
Quantity
Case of 1
Type
Funnel
Volume (Metric)
300 mL
Diameter (Metric) Disc
47 mm