Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass support and GL 45 Style Bottle (47mm)
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass support and GL 45 Style Bottle (47mm)
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass support and GL 45 Style Bottle (47mm)
Designed to collect filtrate directly into a media-style bottle
Have Questions?
Catalog number FISK953750-5347
Price (SGD)
1,946.00
EA
Add to cart
Price (SGD)
1,946.00
EA
Add to cart
Designed to collect filtrate directly into a media-style bottle
  • Supplied with funnel, clamp, fritted glass support base, tubing adapter, PBT cap, sealing ring and vacuum/pressure bottle
  • Support base connects directly to bottle with unique cap and sealing ring
  • Sealed-in 40-60 micron porosity fritted glass disc
  • Vacuum and pressure rated from -1 to 1.5 bar
  • Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
  • All components are steam autoclavable
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
MaterialGlass
TypeMicrofiltration Assembly
Volume (Metric)1 L
Width (English)4 in.
Width (Metric)10.2 cm
Height (English)14.5 in.
Height (Metric)36.8 cm
Unit SizeEA