Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (25mm)
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (25mm)
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (25mm)
Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination
Have Questions?
Catalog number FISK953705-0000
Price (SGD)
813.00
EA
Add to cart
Price (SGD)
813.00
EA
Add to cart
Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination
  • Supplied with a 40-60 micron porosity fritted glass support base, a 15mL graduated funnel, an anodized aluminum clamp and a No. 5 silicone stopper
  • Funnels are graduated from 5 to 15mL in 1mL increments
  • Prefilter size is 16mm diameter
  • Approximate filter area is 2.5cm2
  • Connection to our 125mL filtration flask (available as an accessory) is made with a No. 5 silicone stopper
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
MaterialGlass
Volume (Metric)15 mL
Width (English)1 in.
Width (Metric)2.5 cm
Height (English)6 in.
Height (Metric)15.2 cm
Unit SizeEA