Kimble™ Kontes™ Brand Valved 5mm O.D. NMR Tubes
Kimble™ Kontes™ Brand Valved 5mm O.D. NMR Tubes
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Brand Valved 5mm O.D. NMR Tubes
Ideal for handling volatile, air-sensitive and toxic samples
Have Questions?
Catalog NumberLength (Metric)Camber (Metric)
FISK897435-0000177.8 mm0.013 mm
Catalog number FISK897435-0000
Price (SGD)
610.00
EA
Add to cart
Length (Metric):
177.8 mm
Camber (Metric):
0.013 mm
Price (SGD)
610.00
EA
Add to cart
Ideal for handling volatile, air-sensitive and toxic samples
  • Permit condensing of samples directly into NMR tube
  • Greaseless, Hi-Vac™ valve lets you recover samples, then reseal; no need to permanently seal off, as with conventional tubes
  • With glass tube and Viton™ O-ring for connection to a vacuum system
  • Wall thickness is 0.01475 in. (0.37mm); Wall variation and camber depend, in part, on tube length
Specifications
Camber (Metric)0.013 mm
Diameter (English) Outer0.19 in.
Length (Metric)177.8 mm
MaterialBorosilicate Glass
Diameter (Metric) Outer5 mm
Concentricity (English)0.001 in.
Camber (English)0.0005 in.
Field Strength>360MHz
For Use With (Application)Designed to improve sample handling during the investigation of volatile, air-sensitive, or toxic compounds
Length (English)7 in.
Thickness (English) Wall0.014 in.
Thickness (Metric) Wall0.38 mm
Unit SizeEA