Kimble™ Kontes™ Brand 5mm Disposable Grade NMR Sample Tubes
Kimble™ Kontes™ Brand 5mm Disposable Grade NMR Sample Tubes
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Brand 5mm Disposable Grade NMR Sample Tubes
For instruments with field strength of approximately 60MHz
Have Questions?
Catalog NumberLength (Metric)Quantity
FISK897193-7100177.8 mm100/Pk.
Catalog number FISK897193-7100
Price (SGD)
823.00
CS
Add to cart
Length (Metric):
177.8 mm
Quantity:
100/Pk.
Price (SGD)
823.00
CS
Add to cart
For instruments with field strength of approximately 60MHz
Each tube meets rigid specifications for wall thickness, variation and camber so as to assure precise measurements and reproducibility. Precision-manufactured tubes designed for use in high-resolution NMR spectroscopy that characterizes organic molecules through measurement of the resonant frequencies of atomic nuclei. Research applications include analysis of polymer structures and organic solutions.
  • Made from Type I borosilicate glass
  • Wall thickness: 0.015 in. (0.38mm)
  • Wall variation and camber depend, in part, on tube length
  • All sample tubes come with plastic compression-fit caps and sandblasted marking spots
Specifications
Diameter (English) Outer0.19 in.
Length (Metric)177.8 mm
MaterialBorosilicate Glass
Closure MaterialPolyethylene
Closure TypeOpen-top
Diameter (Metric) Outer5 mm
Field Strength100MHz
For Use With (Application)Designed for use in high-resolution NMR spectroscopy
IncludesCap
Length (English)7 in.
ShapeCylindrical
Thickness (English) Wall0.015 in.
Thickness (Metric) Wall0.38 mm
Quantity100/Pk.
Unit SizeCS