Kimble™ Kontes™ Duall™ Tissue Grinders
Kimble™ Kontes™ Duall™ Tissue Grinders
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Duall™ Tissue Grinders
Combined conical and cylindrical surfaces improve grinding efficiency
Have Questions?
Catalog NumberCapacityIncludesSize
FISK885450-00200.03 oz., 1mLGround Glass pestle and tube20
Catalog number FISK885450-0020
Price (SGD)
583.00
EA
Add to cart
Capacity:
0.03 oz., 1mL
Includes:
Ground Glass pestle and tube
Size:
20
Price (SGD)
583.00
EA
Add to cart
Combined conical and cylindrical surfaces improve grinding efficiency
  • Can be motor driven
  • Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
  • Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance
Specifications
Capacity0.03 oz., 1mL
Capacity (English)0.03 oz.
Diameter (English) Shaft0.23 in.
Diameter (Metric) Shaft6mm
For Use With (Application)Ideal for connective tissue such as muscle, heart and lung
IncludesGround Glass pestle and tube
Length (English) Shaft6.1 in.
Length (English) Tube3.14 in.
Length (Metric) Shaft155mm
Length (Metric) Tube80mm
Size20
Tube Diameter (English) Outside0.51 in.
Tube Diameter (Metric) Outside13mm
Capacity (Metric)1 mL
Clearance (English)0.004 to 0.006 in.
Clearance (Metric)0.1 to 0.15mm
Unit SizeEA