Kimble™ Kontes™ Dounce Tissue Grinders
Kimble™ Kontes™ Dounce Tissue Grinders
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Dounce Tissue Grinders
Designed for cellular work in which the nucleus remains intact after homogenization
Have Questions?
Catalog NumberCapacityClearance (English)Length (English)
FISK885300-00020.06 oz., 2mL0.0005 to 0.0025 in.2.3 in.
Catalog number FISK885300-0002
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
0.06 oz., 2mL
Clearance (English):
0.0005 to 0.0025 in.
Length (English):
2.3 in.
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Specifications
Clearance (English)0.0005 to 0.0025 in.
Clearance (Metric)0.07 to 0.127mm
Capacity (English)0.06 oz.
Capacity (Metric)2 mL
MaterialGlass
Product TypeDounce Tissue Grinder
For Use With (Application)Designed for cellular work in which the nucleus remains intact after homogenization
Length (English)2.3 in.
IncludesOne per case.
Length (Metric)60mm
Capacity0.06 oz., 2mL
Unit SizeEA