Check Order Status
Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
Have Questions?
Change view
Catalog Number
Diameter (Metric)
Standard Taper Joint
FISK774261-0024
2.4 cm
24/40; 24/20
FISK774261-0014
1.4 cm
14/20
2 Options
Catalog number
FISK774261-0024
Price (SGD)
395.00
PK
Add to cart
Diameter (Metric):
2.4 cm
Standard Taper Joint:
24/40; 24/20
Price (SGD)
395.00
PK
Add to cart
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
Diaphragm can be punctured with a syringe needle
Use for outgassing NMR sample tubes, as a pressure vent or for adding reactant via a syringe
Specifications
Diameter (Metric)
2.4 cm
For Use With (Equipment)
NMR tubes, pressure venting or adding reactants via syringe
Standard Taper Joint
24/40; 24/20
Color
White
Material
Rubber
Product Type
Plug-type Sleeve Stopper