Kimble™ Kontes™ Pellet Pestle™
Kimble™ Kontes™ Pellet Pestle™
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Pellet Pestle™
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MaterialIncludesDnase Free
FISK749521-15901.5 mLPolypropylenew/o MicrotubesRNase, DNase and Pyrogen-free
FISK749520-00001.5 mLPolypropylene100 MicrotubesNo
FISK749520-05000.5 mLPolypropylene100 MicrotubesNo
FISK749521-05900.5 mLPolypropylenew/o MicrotubesRNase, DNase and Pyrogen-free
FISK749520-00901.5 mLPolypropylene100 MicrotubesRNase, DNase and Pyrogen-free
FISK749520-05900.5 mLPolypropylene100 MicrotubesRNase, DNase and Pyrogen-free
FISK749521-05000.5 mLPolypropylenew/o MicrotubesNo
FISK749521-15001.5 mLPolypropylenew/o MicrotubesNo
Catalog number FISK749521-1590
Price (SGD)
564.00
PK
Add to cart
Capacity (Metric):
1.5 mL
Material:
Polypropylene
Includes:
w/o Microtubes
Dnase Free:
RNase, DNase and Pyrogen-free
Price (SGD)
564.00
PK
Add to cart
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Disposable Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size
Specifications
Capacity (English)0.05 oz.
Capacity (Metric)1.5 mL
MaterialPolypropylene
Product TypePellet Pestle
DisposableSingle-use
For Use With (Application)Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Length (English)2.75 in.
Dnase FreeRNase, DNase and Pyrogen-free
Includesw/o Microtubes
Length (Metric)70mm
Capacity0.05 oz., 1.5mL
Unit SizePK