Kimble™ Disposable Micro Capillary Pipets
Kimble™ Disposable Micro Capillary Pipets
Actual product may vary
DWK Life Sciences
Kimble™ Disposable Micro Capillary Pipets
Color-coded for selection of correct size
Have Questions?
Catalog NumberVolume (Metric)ColorTolerance
FISK71900-55 μLWhite±1 μL
Catalog number FISK71900-5
Price (SGD)
1,070.00
CS
Add to cart
Volume (Metric):
5 μL
Color:
White
Tolerance:
±1 μL
Price (SGD)
1,070.00
CS
Add to cart
Color-coded for selection of correct size
  • Length: 5 in. with single black capacity ring
  • Clean and ready to use
  • To dispense, simply cover hole on rubber bulb and squeeze
Specifications
Certifications/ComplianceASTM , E672, USP type III
MaterialSoda Lime Glass
Length (English)5 in.
Tolerance±1 μL
ColorWhite
Volume (Metric)5 μL
Length (Metric)127 mm
TypeMicrocapillary Pipet