Kimble™ Kontes™ Wide Mouth Class A Volumetric Flasks
Kimble™ Kontes™ Wide Mouth Class A Volumetric Flasks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Wide Mouth Class A Volumetric Flasks
Specially designed wide mouth mixing bulb for easy access of content in the flask.
Have Questions?
Catalog NumberCapacity (Metric)Length (English)Tolerance
FISK623010-002020 mL125mm±0.08 mL
Catalog number FISK623010-0020
Price (SGD)
245.00
EA
Add to cart
Capacity (Metric):
20 mL
Length (English):
125mm
Tolerance:
±0.08 mL
Price (SGD)
245.00
EA
Add to cart
Specially designed wide mouth mixing bulb for easy access of content in the flask
  • Compatible for applications such as dissolving tablets and other solids
  • Mixing bulb in neck accommodates approximately one tenth of the flask's total volume
  • Capacity range: 5 to 2000mL
Specifications
Capacity (English)0.67 oz.
Capacity (Metric)20 mL
MaterialBorosilicate Glass
Height (English)4.92 in.
Height (Metric)12.5 cm
Length (English)125mm
Neck StyleBulb
No. per CaseCase of 1
Tolerance±0.08 mL
Mouth StyleWide
Closure Size9
Unit SizeEA