Kimble™ Kontes™ Erlenmeyer Flasks
Kimble™ Kontes™ Erlenmeyer Flasks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Erlenmeyer Flasks
Used to measure, mix, and store liquids
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)Standard Taper Joint
FISK617000-0224125 mL12.5 cm24/40
Catalog number FISK617000-0224
Price (SGD)
153.00
EA
Add to cart
Capacity (Metric):
125 mL
Height (Metric):
12.5 cm
Standard Taper Joint:
24/40
Price (SGD)
153.00
EA
Add to cart
Used to measure, mix, and store liquids
  • Single neck flask
  • Has standard taperouter joint
Specifications
Capacity (Metric)125 mL
MaterialBorosilicate Glass
Height (English)4.92 in.
Height (Metric)12.5 cm
Standard Taper Joint24/40
Mouth StyleNarrow
Unit SizeEA