Check Order Status
Kimble™ Kontes™ Round Bottom Three Neck Heavy Wall Distilling Flasks
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Round Bottom Three Neck Heavy Wall Distilling Flasks
With outer joints
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Standard Taper Joint
FISK606000-2324
3000 mL
29/42 Center, 24/40 side
FISK606000-1824
2000 mL
24/40 Center, 24/40 side
2 Options
Catalog number
FISK606000-2324
Price (SGD)
830.00
EA
Add to cart
Capacity (Metric):
3000 mL
Standard Taper Joint:
29/42 Center, 24/40 side
Price (SGD)
830.00
EA
Add to cart
Vertical side necks
Specifications
Diameter (English) Outer
7.28 in.
Capacity (Metric)
3000 mL
Material
Borosilicate Glass
Includes
Standard taper outer joint
Neck Style
Vertical
No. of Positions
3
Standard Taper Joint
29/42 Center, 24/40 side
Bottom Shape
Round
Diameter (Metric) Outer
185 mm