Kimble™ Kontes™ Kuderna-Danish Apparatus with a Graduated Concentrator Tube
Kimble™ Kontes™ Kuderna-Danish Apparatus with a Graduated Concentrator Tube
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Kuderna-Danish Apparatus with a Graduated Concentrator Tube
The Kuderna-Danish apparatus was developed for the concentration of trace amounts of sample dissolved in organic solvents. The column is designed to speed evaporation with reduced hold-up. It is very useful in sample preparation before analysis with solvents such as petroleum ether or hexane.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (English)Height (Metric)Includes
FISK570035-025022.04 in.56 cmHooked model with polyacetal clamps, 250mL flask
FISK570025-050023.62 in.60 cmHooked model with springs, 500mL flask
Catalog number FISK570035-0250
Price (SGD)
1,219.00
EA
Add to cart
Height (English):
22.04 in.
Height (Metric):
56 cm
Includes:
Hooked model with polyacetal clamps, 250mL flask
Price (SGD)
1,219.00
EA
Add to cart
  • Graduated concentrator tube
  • Tube Capacity 10mL
  • Tube Subdivisions (mL): 0-1mL in 0.1mL units, 2-10mL in 1mL units
  • For solvent recovery, a 547300 or 547400 Solvent Recovery Apparatus may be added to this unit
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Preparation involves filling the flask to between 40 to 60 percent of capacity. To prevent sample loss initially, column should be pre-wet with about 1mL of the solvent used in the concentration. If solvent is allowed to escape, entire assembly should be set up in a hood. Charged assembly should be placed over a vigorously boiling water bath. Water level should be maintained just below the lower joint and apparatus mounted so that the lower rounded surface of the flask is bathed in steam. Final sample remains in the lower tube for further analysis.
Specifications
Product TypeGraduated Concentrator Tube
Capacity (Metric)10 mL
Capacity (English)0.3 oz.
Graduations0 to 1 mL/0.1 mL, 2 to 10 mL/1 mL
IncludesHooked model with polyacetal clamps, 250mL flask
For Use With (Application)It is very useful in sample preparation before analysis with solvents such as petroleum ether or hexane
Standard Taper Joint19/22
Standard Taper Joint Top24/40
Height (English)22.04 in.
Height (Metric)56 cm
Unit SizeEA