Kimble™ Kontes™ FlexColumn™ Economy Columns
Kimble™ Kontes™ FlexColumn™ Economy Columns
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ FlexColumn™ Economy Columns
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Column CapacityQuantity
FISK420400-2520200 mm98 mL5/Cs.
FISK420401-2520200 mm98 mLEach
FISK4204002515150 mm74 mL5/Cs.
FKS12973351150 mm74 mL5/Cs.
FISK420400250550 mm25 mL5/Cs.
Catalog number FISK420400-2520
Price (SGD)
825.00
CS
Add to cart
Length (Metric):
200 mm
Column Capacity:
98 mL
Quantity:
5/Cs.
Price (SGD)
825.00
CS
Add to cart
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
  • Durable borosilicate glass
  • With flow adapters
  • Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
  • Threaded, easy to remove reservoir cap won't pop off under pressure
  • Reservoir accepts accessory funnels and glass reservoirs
  • Luer-Lock polypropylene inlet and outlet fittings
  • Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
  • Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Specifications
EndcappedYes
For Use With (Application)Ion exchange, affinity, gel filtration and hydrophobic interaction media
Quantity5/Cs.
Temperature (Metric) Range0 to 50°C
Diameter (Metric) Inner25 mm
TypeEconomy Chromatography Column
Max. Pressure14.5 psi
Pore Size20 μm
Column Capacity98 mL
Length (Metric)200 mm
MaterialBorosilicate Glass Type I Class A
Support MaterialHDPE
Connector TypesMale luer lock
Cross Sectional Area4.91cm3
Sterilization Method2N NaOH or 100% Ethanol
Unit SizeCS