Kimble™ Kontes™ Column with Reservoir
Kimble™ Kontes™ Column with Reservoir
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Column with Reservoir
Reservoir capacities from 200 to 500mL
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) Inner
FISK420280-0213250 mm11 mm
Catalog number FISK420280-0213
Price (SGD)
584.00
EA
Add to cart
Length (Metric):
250 mm
Diameter (Metric) Inner:
11 mm
Price (SGD)
584.00
EA
Add to cart
Reservoir capacities from 200 to 500mL
  • Stopcock has 2mm PTFE plug, threaded pressure nut, and 1:5 taper for easier use at greater-than-ambient temperatures
  • Packing not included
Specifications
For Use With (Application)Analytical separation
Diameter (Metric) Inner11 mm
TypePlain Chromatography Column
Column Capacity200 mL
Length (Metric)250 mm
MaterialGlass
Support MaterialGlass Wool
IncludesIntegral reservoir with size 2 PTFE stopcock plug
Stopper Number2
Unit SizeEA