Check Order Status
Kimble™ Kontes™ FlexColumn™ Three-way Luer Stopcock
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ FlexColumn™ Three-way Luer Stopcock
Heavy-duty nylon
Have Questions?
Catalog number
FISK420163-4503
Price (SGD)
300.00
PK
Add to cart
Price (SGD)
300.00
PK
Add to cart
Heavy-duty nylon
Three-way control with locking fittings on all ports
Specifications
Type
3-way Stopcock
Material
Nylon
For Use With (Equipment)
FlexColumn™
Includes
Locking fittings on all ports
Plug Material
HDPE
Outlet Port
Female Luer to Male Luer
Connectors
Female Luer to Male Luer