Kimble™ Kontes™ Cadium Reduction Chromatography Column
Kimble™ Kontes™ Cadium Reduction Chromatography Column
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Cadium Reduction Chromatography Column
BEVEL-SEAL connections
Have Questions?
Catalog NumberLength (Metric)
FISK419000-8505350 mm
Catalog number FISK419000-8505
Price (SGD)
880.00
CS
Add to cart
Length (Metric):
350 mm
Price (SGD)
880.00
CS
Add to cart
BEVEL-SEAL connections
  • Borosilicate glass
  • 85mL reservoir
  • Glass wool plug
  • PTFE tubing
  • Includes PTFE BEVEL-SEAL stopcock connector
  • 350mm height
Specifications
TypeChromatography Column
Length (Metric)350 mm
MaterialBorosilicate Glass
IncludesReservoir and Bevel-Seal™ stopcock connector
Unit SizeCS