Kimble™ Kontes™ Micro Soxhlet Extraction Apparatus
Kimble™ Kontes™ Micro Soxhlet Extraction Apparatus
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Micro Soxhlet Extraction Apparatus
This Soxhlet-type extraction apparatus is for use with 292100-0000 extraction thimble, which has a coarse porosity.
Have Questions?
Catalog number FISK292010-0000
Price (SGD)
1,292.00
EA
Add to cart
Price (SGD)
1,292.00
EA
Add to cart
  • Capacity 25mL, fits Standard Taper joint 14/20
  • Apparatus requires K292100-0000 extraction thimble, which has a coarse porosity
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Product TypeExtraction Apparatus
For Use With (Equipment)Extraction Thimble K292100-0000
Capacity (Metric)25 mL
Capacity (English)0.84 oz.
IncludesReflux condenser, extractor body and round bottom flask
Standard Taper Joint Condenser19/22
Standard Taper Joint Flask14/20
Height (English)13.38 in.
Height (Metric)34 cm
Unit SizeEA