Kimble™ Kontes™ Bent Drying Tubes with Standard Taper Joints
Kimble™ Kontes™ Bent Drying Tubes with Standard Taper Joints
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Bent Drying Tubes with Standard Taper Joints
Bent at 75° for use in fume hoods where a vertical-type tube won't fit
Have Questions?
Catalog NumberStandard Taper Joint No.
FISK291100-000014/20
Catalog number FISK291100-0000
Price (SGD)
178.00
EA
Add to cart
Standard Taper Joint No.:
14/20
Price (SGD)
178.00
EA
Add to cart
  • With standard taper joint at one end
  • Beaded rim resists breakage
  • Overall H × W: 65 × 105mm
Specifications
Certifications/ComplianceUSP Type I, ASTM E438, Type I, Class A
Dimensions (L x W)2.5 x 4.1 in. (65 x 105 mm)
For Use With (Equipment)Fume hoods
IncludesA 00 rubber stopper with hole to accommodate a 3mm glass inlet tube
Length (English)2.5 in.
Length (Metric)65 mm
ShapeBent
Width (English)4.1 in.
Width (Metric)105 mm
MaterialBorosilicate Glass
Product TypeDrying Tube
Standard Taper Joint No.14/20
Unit SizeEA