Kimble™ KIMAX™ Organic Chemistry Kits
Kimble™ KIMAX™ Organic Chemistry Kits
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Organic Chemistry Kits
Include an assortment of items required for basic organic laboratory experiments
Have Questions?
Catalog NumberProduct TypeStandard Taper Joint
FISK269800-0000Standard Taper Kit19/22
Catalog number FISK269800-0000
Price (SGD)
2,679.00
EA
Add to cart
Product Type:
Standard Taper Kit
Standard Taper Joint:
19/22
Price (SGD)
2,679.00
EA
Add to cart
  • For education and research
  • All joints in a kit are the same standard taper size, making it easy to set up a number of procedures
Specifications
Certifications/ComplianceUSP Type I and ASTM E438 Type I, Class A
Product TypeStandard Taper Kit
For Use With (Application)For extraction, reflux, reflux with addition, fractionation, and distillation
IncludesBleed adapter, claisen adapter, condenser, connecting adapter, distillation column, 25mL, 50mL, 100mL, 250mL, 500mL Flask, 125mL PTFE funnel, outlet adapter, stopper, vacuum adapter
Standard Taper Joint19/22
Unit SizeEA