Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Glassware
▸
Arsine Glassware
▸
Kimble™ Kontes™ Arsine Generator Absorber Tube
Actual product may vary
DWK Life Sciences
Kimble™ Kontes™ Arsine Generator Absorber Tube
This Arsine Generator Absorber Tube is a Component Part For Arsine Generators
Have Questions?
Catalog Number
Standard Taper Joint
FISK2411020000
12/2
Catalog number
FISK2411020000
Price (SGD)
202.00
Each
Add to cart
Standard Taper Joint:
12/2
Price (SGD)
202.00
Each
Add to cart
Arsine Generator Absorber Tube Only
Spherical Joint 12/2
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Specifications
Product Type
Absorber Tube
Standard Taper Joint
12/2
For Use With (Equipment)
For arsine generator