Durapore™ Membrane Filters 0.45 μm
Durapore™ Membrane Filters 0.45 μm
Actual product may vary
MilliporeSigma
Durapore™ Membrane Filters 0.45 μm
Recommended when maximum recovery is important
Have Questions?
Catalog NumberDiameter (Metric)Surface Type
FISHVWG0470047 mmGridded
Catalog number FISHVWG04700
Price (SGD)
808.00
PK
Add to cart
Diameter (Metric):
47 mm
Surface Type:
Gridded
Price (SGD)
808.00
PK
Add to cart
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.45μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Durapore™ membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic Durapore™ membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.
    Features & Benefits
  • Available in several pore sizes (both hydrophilic and hydrophobic varieties) to suit your application needs
  • Durapore™ membrane filters have very low protein binding to minimize interaction with your sample and maximize recovery
  • Specifications
    AutoclavableNot Autoclavable
    Bubble Point22 psi
    ColorWhite
    Filter TypeScreen Filter
    MaterialPVDF
    Pore Size0.45 μm
    SterilityNon-sterile
    Flow Rate>2.6 mL/min x cm2
    HydrophobicityHydrophilic
    No. per Case100Pack
    Quantity100/Pk.
    Surface TypeGridded
    Thickness (Metric)125 μm
    TypeFilter Membranes
    WettabilityHydrophilic
    Diameter (Metric)47 mm
    Unit SizePK