Durapore™ Membrane Filters, 0.22 μm
Durapore™ Membrane Filters, 0.22 μm
Actual product may vary
MilliporeSigma
Durapore™ Membrane Filters, 0.22 μm
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Have Questions?
Catalog NumberDiameter (Metric)DimensionsLxWWettability
FISGVWP0470047 mmHydrophilic
Catalog number FISGVWP04700
Price (SGD)
689.00
PK
Add to cart
Diameter (Metric):
47 mm
Wettability:
Hydrophilic
Price (SGD)
689.00
PK
Add to cart
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Durapore™ membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic Durapore™ membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.
    Features & Benefits
  • Available in several pore sizes (both hydrophilic and hydrophobic varieties) to suit your application needs
  • Durapore™ membrane filters have very low protein binding to minimize interaction with your sample and maximize recovery
  • Specifications
    AutoclavableNot Autoclavable
    Bubble Point50 psi
    ColorWhite
    Filter TypeScreen Filter
    MaterialPVDF
    Pore Size0.22 μm
    SterilityNon-sterile
    Flow Rate>1 mL/min x cm2
    Gas Flow Rate2 L/min/ cm2
    HydrophobicityHydrophilic
    Membrane TypePlain
    No. per Case100Pack
    Quantity100/Pk.
    Thickness (Metric)125 μm
    TypeFilter Membranes
    WettabilityHydrophilic
    Diameter (Metric)47 mm
    Unit SizePK