dNTP Mix (25 mM each)
dNTP Mix (25 mM each)
Actual product may vary
Fisher Catalogue
dNTP Mix (25 mM each)
Thermo Scientific dNTP Mix (25 mM) contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 25 mM.
Have Questions?
Catalog NumberQuantity
FISFERR11211 mL
Catalog number FISFERR1121
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mL
Thermo Scientific dNTP Mix (25 mM) contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 25 mM. The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. Mixes offer the possibility to reduce the number of pipetting steps and the risk of reaction set up errors. They are designed for many different molecular biology applications.
Features
• Greater than 99% purity confirmed by HPLC
• Free of human and E. coli DNA
• Long term stability, even after multiple freeze-thaw cycles when properly stored
• Up to 95% of dNTPs remain in triphosphate form after 7 weeks at room temperature
• Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 minute at 94°C, 3 minutes at 72°C)
Applications
• Long range PCR (40 kb)
• cDNA synthesis
• PCR, RT-PCR, qPCR
• High fidelity PCR
• DNA labeling and sequencing
• LAMP, RDA, MDA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Label or DyeUnlabeled
Purity or Quality Grade>99%
Quantity1 mL
Shipping ConditionDry Ice
For Use With (Application)Standard PCR, High-fidelity PCR, RT-PCR, Real Time PCR (qPCR)
Concentration25 mM
Size (Final Product)40 kb
Unit SizeEA
Contents & Storage
dNTP Mix (25 mM) contains premixed dATP, dCTP, dGTP and dTTP at a final concentration of 25 mM (1 mL)
Store at –20°C.