Sterile PETG Media Bottles
Actual product may vary
Sterile PETG Media Bottles
Sterile PETG Media Bottles
Sterile PETG Media Bottles
Fisherbrand
Sterile PETG Media Bottles
Reduce the risk of cross contamination with these sterile, disposable media bottles
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Type
FISFBMB6060 mLTamper-evident Screw Cap
FISFBMB3030 mLTamper-evident Screw Cap
FISFBMB60A60 mLStandard Screw Cap
FISFBMB125A125 mLStandard Screw Cap
FISFBMB125125 mLTamper-evident Screw Cap
FISFBMB250250 mLTamper-evident Screw Cap
FISFBMB500500 mLTamper-evident Screw Cap
FISFBMB1000A1000 mLStandard Screw Cap
FISFBMB10001000 mLTamper-evident Screw Cap
Catalog number FISFBMB60
Price (SGD)
392.00
Case
Capacity (Metric):
60 mL
Closure Type:
Tamper-evident Screw Cap
Price (SGD)
392.00
Case
Reduce the risk of cross contamination with these sterile, disposable media bottles
The Fisherbrand PETG Media Bottle is ideal for media preparation and storage.
  • An inexpensive and shatterproof alternative to glass media bottles
  • Nonpyrogenic and noncytotoxic to protect cells
  • Molded-in graduations won't rub off
  • Made from clear, lightweight, polyethylene terephthalate copolyester (PETG) providing impact resistance and excellent gas-barrier properties
  • Closure is is high density polyethylene (HDPE)
  • Bottles packed in a shrink-wrapped tray to stay clean
  • Heat-shrink band around cap and bottle neck provides visual assurance of sterility for individual bottles until band is removed
  • Part numbers ending in "A" are designed without a temper-evident ring
Specifications
Capacity2.02 oz., 60mL
Capacity (Metric)60 mL
ColorClear
For Use With (Application)For media preparation and storage
MaterialPETG
SterilitySterile
Closure MaterialHDPE
Closure Size24 mm
Closure TypeTamper-evident Screw Cap
Capacity (English)2.02 oz.
Certifications/ComplianceSAL 10-6, USP Class VI
Product TypeMedia Bottle
Unit SizeCase