Check Order Status
Mortar and Pestle Regular Form
Actual product may vary
Fisherbrand
Mortar and Pestle Regular Form
Mortar and Pestle Regular Form
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Height (Metric)
FISFB970C
9 mL
28mm
FISFB970B
6 mL
25mm
2 Options
Catalog number
FISFB970C
Price (SGD)
161.00
EA
Add to cart
Capacity (Metric):
9 mL
Height (Metric):
28mm
Price (SGD)
161.00
EA
Add to cart
Low permeability
High strength
Hard
High resistance to chemical attacks
Translucent or glazed body
Specifications
Capacity (Metric)
9 mL
Diameter (Metric) Inner
38mm
Diameter (Metric) Outer
50mm
Product Type
Mortar and Pestle
Material
Agate Stone
For Use With (Application)
For grinding or crushing
Height (Metric)
28mm
Includes
Mortar and Pestle