Porcelain Gooch Filtering Crucibles
Porcelain Gooch Filtering Crucibles
Actual product may vary
Porcelain Gooch Filtering Crucibles
For total suspended solids determinations
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)Thickness (Metric)
FISFB969B40 mL41mm51mm2.2mm
FISFB969A25 mL36mm45mm2mm
Catalog number FISFB969B
Price (SGD)
198.00
PK
Add to cart
Capacity (Metric):
40 mL
Diameter (Metric) Outer:
41mm
Height (Metric):
51mm
Thickness (Metric):
2.2mm
Price (SGD)
198.00
PK
Add to cart
Glazed on the outside, except for bottom surface and rim
  • Perforated bottom
  • Withstand temperatures up to 1150°C (2102°F)
  • Without cover
Specifications
DescriptionTolerance: ±5%
Diameter (Metric) Inner25mm
MaterialPorcelain
For Use With (Application)For total suspended solids determination
Max. Temperature (Metric)1150°C
Thickness (Metric)2.2mm
Capacity (Metric)40 mL
Diameter (Metric) Outer41mm
Height (Metric)51mm
Product TypeGooch Filtering Crucible
Unit SizePK