Check Order Status
Porcelain Evaporating Dishes
Actual product may vary
Porcelain Evaporating Dishes
Glazed inside and out except for rim.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FISFB968F
250 mL
FISFB968C
80 mL
FISFB968D
120 mL
FISFB962K
50 mL
4 Options
Catalog number
FISFB968F
Price (SGD)
391.00
PK
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
391.00
PK
Add to cart
Porcelain dish with lip
Glazed inside and out except for rim
Highly resistant to chemicals
Withstand temperatures up to 1150°C (2102°F)
Without cover
Specifications
Bottom Shape
Round
Capacity (Metric)
250 mL
Material
Porcelain
Shape
Round
Type
Evaporating Dish
For Use With (Application)
Evaporating Application
Interior Finish
Glazed
Thickness
3 mm
Height (Metric)
45 mm
Height (English)
1.77 in.
Diameter (English)
4.52 in.
Diameter (Metric)
115 mm