Porcelain Crucible Covers
Porcelain Crucible Covers
Actual product may vary
Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) OuterDiameter (Metric) InnerHeight (Metric)Thickness (Metric)
FISFB965X87mm77mm8mm2.75mm
FISFB965U53mm45mm7mm2.3mm
FISFB965W75mm66mm7mm2.5mm
FISFB965V60mm54mm7mm2.5mm
Catalog number FISFB965X
Price (SGD)
41.00
PK
Add to cart
Diameter (Metric) Outer:
87mm
Diameter (Metric) Inner:
77mm
Height (Metric):
8mm
Thickness (Metric):
2.75mm
Price (SGD)
41.00
PK
Add to cart
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
DescriptionWith Perforated handle
Diameter (Metric) Inner77mm
MaterialPorcelain
FinishGlazed
For Use With (Application)Use with Fisherbrand high and wide form porcelain crucible
Max. Temperature (Metric)1150°C
Thickness (Metric)2.75mm
Weight (Metric)56g
Diameter (Metric) Outer87mm
Height (Metric)8mm
Product TypeCrucible Cover
Unit SizePK