Porcelain Combustion Boats
Actual product may vary
Porcelain Combustion Boats
Chemical porcelain labware design with resistance to heat and caustic chemicals
Have Questions?
Catalog NumberDimensions (L x W x H)
FISFB963D100 x 20 x 13 mm
Catalog number FISFB963D
Price (SGD)
63.00
Each
Dimensions (L x W x H):
100 x 20 x 13 mm
Price (SGD)
63.00
Each
Chemical porcelain labware design with resistance to heat and caustic chemicals
  • Porcelain made
  • Color: White
  • Maximum use temperature is 1150° with a heating & cooling rate not exceeding 200°C per hour
The heating and cooling cycles can actually withstand a more rigorous heating rate than recommended. However, excess of thermal shock can damage these products.
Specifications
TypeCombustion Boat
ColorWhite
Dimensions (L x W x H)100 x 20 x 13 mm
For Use With (Application)Scientific tools used in experiments because of their resistance to heat and caustic chemicals
MaterialPorcelain
Unit SizeEach