Porcelain Pestles
Porcelain Pestles
Actual product may vary
Porcelain Pestles
For hand comfort in grinding
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Length (English)Length (Metric)
FISFB961P750mL mortar8.7 in.22.2cm
FISFB961L65mL mortar5.1 in.13cm
FISFB961K50mL mortar4.49 in.11.4cm
FISFB961O400mL Mortar7.6 in.19.4cm
FISFB961N275mL Mortar7 in.18cm
Catalog number FISFB961P
Price (SGD)
196.00
EA
Add to cart
For Use With (Equipment):
750mL mortar
Length (English):
8.7 in.
Length (Metric):
22.2cm
Price (SGD)
196.00
EA
Add to cart
For hand comfort in grinding
  • Unglazed on grinding surface
  • Autoclavable
  • Oversized
Specifications
FinishUnglazed Grinding Surface
AutoclavableAutoclavable
Product TypePestle
MaterialPorcelain
For Use With (Equipment)750mL mortar
For Use With (Application)For hand comfort in grinding
Length (Metric)22.2cm
Length (English)8.7 in.
Unit SizeEA