▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
Crucibles
▸
Alumina Crucibles
Actual product may vary
Alumina Crucibles
Made up of 99.7% Alumina
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Product Type
FISFB960A
2 mL
Cylindrical form crucible
FISFB960I
10 mL
High-Form Crucible
FISFB960K
50 mL
High-Form Crucible
3 Options
Catalog number
FISFB960A
Price (SGD)
-
Capacity (Metric):
2 mL
Product Type:
Cylindrical form crucible
Made up of 99.7% Alumina
Available in two forms: high and cyclindrical
Unglazed
Maximum use temperature: 1750°C
Resistant to Alkalies, chemical attack and other fluxes
Inert in hydrogen and carbonaceous atmospheres
Color: cream
Specifications
Color
Cream
Material
Alumina
Max. Temperature (Metric)
1750°C
Capacity (Metric)
2 mL
Height (Metric)
25mm
Product Type
Cylindrical form crucible
Shape
Cylinder