High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
Actual product may vary
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
High-Form Alumina Crucibles Covers
Covers for High-Form Alumina crucibles
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
FISFB9603For 50mL crucible
FISFB9601For 10mL crucible
Catalog number FISFB9603
Price (SGD)
240.00
PK
Add to cart
For Use With (Application):
For 50mL crucible
Price (SGD)
240.00
PK
Add to cart
  • For 0.3, 0.6, 1.6 and 3.3 oz. (10, 20, 50 and 100mL) crucibles
  • Alumina construction
  • Color: cream
Specifications
ColorCream
MaterialAlumina
For Use With (Application)For 50mL crucible
Product TypeCrucible Cover
Unit SizePK