Check Order Status
Replacement Cap for Glass Media Bottles
Actual product may vary
Replacement Cap for Glass Media Bottles
Fits GL45 screw thread
Have Questions?
Catalog Number
Thread Size (Metric)
FISFB800STDCAP
GL 45
Catalog number
FISFB800STDCAP
Price (SGD)
116.00
CS
Add to cart
Thread Size (Metric):
GL 45
Price (SGD)
116.00
CS
Add to cart
This Fisherbrand™ Replacement Cap fits GL45 screw thread
Autoclavable up to 140°C
Polypropylene
Linerless Blue
Specifications
Color
Blue
Material
Polypropylene
No. per Case
10 Cs.
Sterility
Non-sterile
Thread Size (Metric)
GL 45
Autoclavable
Autoclavable
Product Type
Screw Cap
For Use With (Equipment)
Media bottle