Reusable Glass Class A Volumetric Flasks with Snap Caps
Reusable Glass Class A Volumetric Flasks with Snap Caps
Actual product may vary
Reusable Glass Class A Volumetric Flasks with Snap Caps
Class A accuracy–without a serial number
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FISFB40120002000 mL8
FISFB4011010 mL2
FIS10-200B50 mL16
FIS10-200D200 mL16
FIS10-200C100 mL16
FIS10-200E250 mL16
FIS10-200F500 mL16
FIS10-200G1000 mL16
Catalog number FISFB4012000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2000 mL
Closure Size:
8
Class A accuracy–without a serial number
  • Calibrated “To Contain”
Specifications
CalibrationCalibrated To Contain
Closure MaterialPolyethylene
Capacity (Metric)2000 mL
MaterialGlass
Height (English)14.5 in.
Height (Metric)37 cm
No. per Case1
GraduatedYes
Closure Size8
Unit SizeEA