Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size |
---|---|---|
FISFB4002000 | 2000 mL | 27 |
FISFB40010 | 10 mL | 9 |
FISFB400200 | 200 mL | 16 |
FIS10-205A | 25 mL | 9 |
FIS10-205B | 50 mL | 9 |
FIS10-205C | 100 mL | 13 |
FIS10-205D | 250 mL | 16 |
FIS10-205E | 500 mL | 19 |
FIS10-205F | 1000 mL | 22 |