Reusable Heavy-Wall Filter Flasks
Reusable Heavy-Wall Filter Flasks
Actual product may vary
Reusable Heavy-Wall Filter Flasks
With heavier walls than standard Erlenmeyer flasks
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Graduation RangeClosure Size
FISFB30020002000 mL30 cm600 to 2000 mL9
FISFB30010001000 mL23.8 cm300 to 1000 mL8
FISFB300250250 mL16.3 cm50 to 250 mL6
FISFB300500500 mL19 cm150 to 500 mL7
Catalog number FISFB3002000
Price (SGD)
215.00
Each
Add to cart
Capacity (Metric):
2000 mL
Height (Metric):
30 cm
Graduation Range:
600 to 2000 mL
Closure Size:
9
Price (SGD)
215.00
Each
Add to cart
With heavier walls than standard Erlenmeyer flasks
  • Side tubulation Capacity scale
  • Designed to take 1 atmosphere of vacuum or 29 in. of mercury
  • Sizes 25 through 500mL have side hose connection designed to accept 5.16 in. O.D. flexible tubing
  • Sizes 1000mL and larger accept 3.8 in. O.D. tubing
  • Temperature range: 840°F (450°C) max.
Specifications
Certifications/ComplianceASTM E 1406, Type II, Class I
Diameter (English) Outer6.6 in.
Diameter (Metric) Inner Tubing0.96 cm
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
Graduation Range600 to 2000 mL
Graduations100 mL
Height (English)11.8 in.
Height (Metric)30 cm
No. per Case1
Temperature Range (English)840°F max.
GraduatedYes
Diameter (Metric) Outer16.8 cm
Closure Size9
Unit SizeEach