Reusable Glass Round-Bottom Boiling Flasks with standard taper Joint
Reusable Glass Round-Bottom Boiling Flasks with standard taper Joint
Actual product may vary
Reusable Glass Round-Bottom Boiling Flasks with standard taper Joint
Feature full length 24/40 standard taper Joint
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Outer
FISFB201500500 mL16 cm102 mm
FISFB201100100 mL11.5 cm63 mm
Catalog number FISFB201500
Price (SGD)
153.00
PK
Add to cart
Capacity (Metric):
500 mL
Height (Metric):
16 cm
Diameter (Metric) Outer:
102 mm
Price (SGD)
153.00
PK
Add to cart
Feature full length 24/40 standard taper Joint
Specifications
Diameter (English) Outer4 in.
Capacity (Metric)500 mL
MaterialGlass
Height (English)6.2 in.
Height (Metric)16 cm
No. per Case1
Standard Taper Joint24/40
DisposableReusable
ClassClass III
Bottom ShapeRound
Diameter (Metric) Outer102 mm
Unit SizePK