Check Order Status
Reusable Glass Flat-Bottom Boiling Flasks with standard taper
Actual product may vary
Reusable Glass Flat-Bottom Boiling Flasks with standard taper
Rugged short standard taper neck
Have Questions?
Catalog Number
Capacity (Metric)
Height (Metric)
Diameter (Metric) Outer
FISFB200250
250 mL
13.3 cm
83 mm
Catalog number
FISFB200250
Price (SGD)
71.00
EA
Add to cart
Capacity (Metric):
250 mL
Height (Metric):
13.3 cm
Diameter (Metric) Outer:
83 mm
Price (SGD)
71.00
EA
Add to cart
Rugged short standard taper neck
Features short 24/40 standard taper
Specifications
Diameter (English) Outer
3.26 in.
Capacity (Metric)
250 mL
Material
Borosilicate Glass
Height (English)
5.23 in.
Height (Metric)
13.3 cm
Neck Style
Short
No. per Case
1
Standard Taper Joint
24/40
Disposable
Reusable
Bottom Shape
Flat
Diameter (Metric) Outer
83 mm